Assay Details
| Gene Symbol: TAS1R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122280.2 | XP_014977766.2 | 638 | 76 | ||
| GenBank mRNA | JU494572.1 | - | 308 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS1R3| Gene Symbol: TAS1R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122280.2 | XP_014977766.2 | 638 | 76 | ||
| GenBank mRNA | JU494572.1 | - | 308 | 76 | ||
Set Membership: |
|