Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091148.4 | XP_001091148.3 | 1116 | 84 | ||
| XM_015126826.2 | XP_014982312.1 | 1094 | 84 | |||
| GenBank mRNA | JU325623.1 | - | 806 | 84 | ||
| JV668364.1 | - | 1068 | 84 | |||
| Gene Symbol: FANCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU325622.1 | - | 806 | 84 | ||
| JU474667.1 | - | 5548 | 84 | |||
| JU474668.1 | - | 5620 | 84 | |||
| JU474670.1 | - | 5548 | 84 | |||
More Information
Set Membership: |
|

ZNF276
Genomic Map