Assay Details
| Gene Symbol: ERP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090991.4 | XP_001090991.2 | 1891 | 100 | ||
| XM_015151200.2 | XP_015006686.2 | 1025 | 100 | |||
| GenBank mRNA | JU479973.1 | - | 665 | 100 | ||
More Information
Set Membership: |
|

ERP27
Genomic Map