Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194401.2 | NP_001181330.1 | 985 | 86 | ||
| XM_015143930.2 | XP_014999416.1 | 1045 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RDH12| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194401.2 | NP_001181330.1 | 985 | 86 | ||
| XM_015143930.2 | XP_014999416.1 | 1045 | 86 | |||
Set Membership: |
|