Assay Details
| Gene Symbol: LOC702140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110660.2 | XP_014966146.2 | 762 | 164 | ||
| XM_015110661.2 | XP_014966147.2 | 761 | 164 | |||
More Information
Set Membership: |
|

LOC702140
Genomic Map