Assay Details
| Gene Symbol: C14H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194273.2 | NP_001181202.1 | 136 | 103 | ||
| XM_015115597.2 | XP_014971083.1 | 128 | 103 | |||
| XM_028832653.1 | XP_028688486.1 | 128 | 103 | |||
| XM_028832654.1 | XP_028688487.1 | 128 | 103 | |||
More Information
Set Membership: |
|

C14H11orf65
Genomic Map