Assay Details
| Gene Symbol: C14H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194273.2 | NP_001181202.1 | 800 | 87 | ||
| XM_015115597.2 | XP_014971083.1 | 738 | 87 | |||
| XM_015115599.2 | XP_014971085.1 | 707 | 87 | |||
| XM_015115601.2 | XP_014971087.1 | 653 | 87 | |||
More Information
Set Membership: |
|

C14H11orf65
Genomic Map