Assay Details
| Gene Symbol: LMNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151315.2 | XP_015006801.1 | 961 | 115 | ||
| XM_015151316.2 | XP_015006802.1 | 813 | 115 | |||
More Information
Set Membership: |
|

LMNTD1
Genomic Map