Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194201.2 | NP_001181130.1 | 372 | 78 | ||
| GenBank mRNA | JV635525.1 | - | 501 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP8| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194201.2 | NP_001181130.1 | 372 | 78 | ||
| GenBank mRNA | JV635525.1 | - | 501 | 78 | ||
Set Membership: |
|