Assay Details
Gene Symbol: THAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194201.2 | NP_001181130.1 | 766 | 103 | ||
GenBank mRNA | JU325282.1 | - | 85 | 103 | ||
JV635525.1 | - | 895 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
