Assay Details
| Gene Symbol: RWDD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257574.1 | NP_001244503.1 | 289 | 142 | ||
| XM_015139574.2 | XP_014995060.1 | 358 | 142 | |||
| GenBank mRNA | JU333319.1 | - | 289 | 142 | ||
| JU474629.1 | - | 272 | 142 | |||
| JV046409.1 | - | 283 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RWDD4
Genomic Map