Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102600.4 | XP_001102600.1 | 260 | 118 | ||
| XM_015130865.2 | XP_014986351.1 | 287 | 118 | |||
| XM_028844156.1 | XP_028699989.1 | 386 | 118 | |||
| XM_028844158.1 | XP_028699991.1 | 293 | 118 | |||
More Information
Set Membership: |
|

LSMEM2
Genomic Map