Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084253.4 | XP_001084253.1 | 310 | 159 | ||
| XM_001084373.4 | XP_001084373.2 | 592 | 159 | |||
| GenBank mRNA | JU323298.1 | - | 260 | 159 | ||
More Information
Set Membership: |
|

IKBIP
Genomic Map