Assay Details
| Gene Symbol: PRXL2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106503.4 | XP_001106503.1 | 580 | 129 | ||
| GenBank mRNA | JU326556.1 | - | 493 | 129 | ||
| JV708795.1 | - | 503 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRXL2C
Genomic Map