Assay Details
| Gene Symbol: POGLUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104429.4 | XP_001104429.1 | 494 | 68 | ||
| XM_015115618.2 | XP_014971104.1 | 453 | 68 | |||
| XM_028833944.1 | XP_028689777.1 | 408 | 68 | |||
More Information
Set Membership: |
|

POGLUT3
Genomic Map