Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145374.2 | XP_015000860.1 | 986 | 149 | ||
| XM_015145376.2 | XP_015000862.1 | 750 | 149 | |||
| XM_028852673.1 | XP_028708506.1 | 1025 | 149 | |||
| XM_028852674.1 | XP_028708507.1 | 614 | 149 | |||
| XM_028852675.1 | XP_028708508.1 | 590 | 149 | |||
| GenBank mRNA | JU325901.1 | - | 489 | 149 | ||
| JV631912.1 | - | 796 | 149 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS20
Genomic Map