Assay Details
Gene Symbol: LOC710876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142078.2 | XP_014997564.2 | 337 | 119 | ||
XM_015142087.2 | XP_014997573.1 | 337 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015142078.2, XM_015142087.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
