Assay Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151789.2 | XP_015007275.2 | 800 | 139 | ||
| XM_015151790.2 | XP_015007276.2 | 424 | 139 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT78| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151789.2 | XP_015007275.2 | 800 | 139 | ||
| XM_015151790.2 | XP_015007276.2 | 424 | 139 | |||
Set Membership: |
|