Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849580.1 | XP_028705413.1 | 1519 | 132 | ||
| GenBank mRNA | JV713489.1 | - | 186 | 132 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFESD| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849580.1 | XP_028705413.1 | 1519 | 132 | ||
| GenBank mRNA | JV713489.1 | - | 186 | 132 | ||
Set Membership: |
|