Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837032.1 | XP_028692865.1 | 1139 | 94 | ||
| XM_028837033.1 | XP_028692866.1 | 702 | 94 | |||
| XM_028837034.1 | XP_028692867.1 | 636 | 94 | |||
| XM_028837035.1 | XP_028692868.1 | 867 | 94 | |||
| XM_028837036.1 | XP_028692869.1 | 643 | 94 | |||

HEXD
Genomic Map