Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194706.3 | NP_001181635.2 | 259 | 126 | ||
| XM_015124423.2 | XP_014979909.2 | 210 | 126 | |||
| XM_015124424.2 | XP_014979910.2 | 307 | 126 | |||
| XM_015124426.2 | XP_014979912.2 | 288 | 126 | |||
| XM_028838533.1 | XP_028694366.1 | 247 | 126 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194706.3, XM_015124423.2, XM_015124424.2, XM_015124426.2, XM_028838533.1 transcripts.
Set Membership: |
|

SIGLECL1
Genomic Map