Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194706.3 | NP_001181635.2 | 542 | 119 | ||
| XM_015124423.2 | XP_014979909.2 | 493 | 119 | |||
| XM_015124424.2 | XP_014979910.2 | 590 | 119 | |||
| XM_015124426.2 | XP_014979912.2 | 571 | 119 | |||
| XM_028838533.1 | XP_028694366.1 | 530 | 119 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SIGLECL1
Genomic Map