Assay Details
| Gene Symbol: RIMKLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261229.1 | NP_001248158.1 | 704 | 87 | ||
| GenBank mRNA | JU334092.1 | - | 704 | 87 | ||
| JU334093.1 | - | 704 | 87 | |||
More Information
Set Membership: |
|

RIMKLA
Genomic Map