Assay Details
| Gene Symbol: ATG9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846476.1 | XP_028702309.1 | 3509 | 112 | ||
| GenBank mRNA | JV465797.1 | - | 57 | 112 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATG9B| Gene Symbol: ATG9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846476.1 | XP_028702309.1 | 3509 | 112 | ||
| GenBank mRNA | JV465797.1 | - | 57 | 112 | ||
Set Membership: |
|