Assay Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148280.2 | XP_015003766.1 | 1360 | 128 | ||
| GenBank mRNA | JU326598.1 | - | 856 | 128 | ||
| JU470953.1 | - | 1147 | 128 | |||
| JV706551.1 | - | 877 | 128 | |||
More Information
Set Membership: |
|

PDZD8
Genomic Map