Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193990.1 | NP_001180919.1 | 251 | 115 | ||
| XM_015143103.1 | XP_014998589.1 | 337 | 115 | |||
| XM_015143104.2 | XP_014998590.1 | 358 | 115 | |||
| XM_015143105.1 | XP_014998591.1 | 361 | 115 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TPPP2
Genomic Map