Assay Details
| Gene Symbol: FAAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095907.4 | XP_001095907.1 | 657 | 104 | ||
| XM_015127557.2 | XP_014983043.1 | 782 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAAH2| Gene Symbol: FAAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095907.4 | XP_001095907.1 | 657 | 104 | ||
| XM_015127557.2 | XP_014983043.1 | 782 | 104 | |||
Set Membership: |
|