Assay Details
| Gene Symbol: TCERG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827020.1 | XP_028682853.1 | 1111 | 100 | ||
| XM_028827021.1 | XP_028682854.1 | 1111 | 100 | |||
| GenBank mRNA | JV463501.1 | - | 165 | 100 | ||
| JV539977.1 | - | 365 | 100 | |||
More Information
Set Membership: |
|

TCERG1L
Genomic Map