Assay Details
| Gene Symbol: CD163L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150972.2 | XP_015006458.2 | 692 | 66 | ||
| XM_028829233.1 | XP_028685066.1 | 598 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD163L1| Gene Symbol: CD163L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150972.2 | XP_015006458.2 | 692 | 66 | ||
| XM_028829233.1 | XP_028685066.1 | 598 | 66 | |||
Set Membership: |
|