Assay Details
| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114986.2 | XP_014970472.1 | 1212 | 135 | ||
| GenBank mRNA | JV597322.1 | - | 787 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCKBR| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114986.2 | XP_014970472.1 | 1212 | 135 | ||
| GenBank mRNA | JV597322.1 | - | 787 | 135 | ||
Set Membership: |
|