Assay Details
| Gene Symbol: SYT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113600.2 | XP_014969086.2 | 1457 | 87 | ||
| XM_028833180.1 | XP_028689013.1 | 1245 | 87 | |||
| XM_028833181.1 | XP_028689014.1 | 1258 | 87 | |||
More Information
Set Membership: |
|

SYT12
Genomic Map