Assay Details
| Gene Symbol: TUBB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092923.4 | XP_001092923.1 | 1586 | 89 | ||
| GenBank mRNA | JV615430.1 | - | 315 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBB2B| Gene Symbol: TUBB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092923.4 | XP_001092923.1 | 1586 | 89 | ||
| GenBank mRNA | JV615430.1 | - | 315 | 89 | ||
Set Membership: |
|