Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110857.4 | XP_001110857.1 | 1296 | 142 | ||
XM_028845672.1 | XP_028701505.1 | 1293 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
