Assay Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194729.2 | NP_001181658.1 | 77 | 142 | ||
| XM_015136276.2 | XP_014991762.1 | 2085 | 142 | |||
| GenBank mRNA | JU323882.1 | - | 74 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194729.2, XM_015136276.2 transcripts.
Set Membership: |
|

TREML1
Genomic Map