Assay Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194729.2 | NP_001181658.1 | 662 | 133 | ||
| GenBank mRNA | JU323882.1 | - | 659 | 133 | ||
| JV682544.1 | - | 309 | 133 | |||
More Information
Set Membership: |
|

TREML1
Genomic Map