Assay Details
| Gene Symbol: NMNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132009.2 | XP_014987495.1 | 652 | 140 | ||
| XM_015132010.2 | XP_014987496.1 | 643 | 140 | |||
| XM_028843745.1 | XP_028699578.1 | 785 | 140 | |||
| XM_028843746.1 | XP_028699579.1 | 731 | 140 | |||
More Information
Set Membership: |
|

NMNAT3
Genomic Map