Assay Details
| Gene Symbol: DRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148115.2 | XP_015003601.1 | 997 | 151 | ||
| XM_015148122.2 | XP_015003608.1 | 1063 | 151 | |||
| XM_015148131.2 | XP_015003617.1 | 1062 | 151 | |||
| XM_015148144.2 | XP_015003630.1 | 1100 | 151 | |||
| XM_028830462.1 | XP_028686295.1 | 1012 | 151 | |||
| XM_028830468.1 | XP_028686301.1 | 1103 | 151 | |||
| XM_028830477.1 | XP_028686310.1 | 1011 | 151 | |||
| XM_028830485.1 | XP_028686318.1 | 879 | 151 | |||
| XM_028830488.1 | XP_028686321.1 | 739 | 151 | |||
| XM_028830490.1 | XP_028686323.1 | 916 | 151 | |||
| XM_028830493.1 | XP_028686326.1 | 871 | 151 | |||
| GenBank mRNA | JU326789.1 | - | 600 | 151 | ||
| JU474319.1 | - | 689 | 151 | |||
| JV046192.1 | - | 689 | 151 | |||
More Information
Set Membership: |
|

DRAM2
Genomic Map