Assay Details
| Gene Symbol: MFSD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093314.4 | XP_001093314.2 | 1453 | 83 | ||
| GenBank mRNA | JU333086.1 | - | 1419 | 83 | ||
| JU333087.1 | - | 1419 | 83 | |||
| JV606847.1 | - | 1456 | 83 | |||
More Information
Set Membership: |
|

MFSD4A
Genomic Map