Assay Details
| Gene Symbol: GPR141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133696.2 | XP_014989182.2 | 5758 | 132 | ||
| XM_028845861.1 | XP_028701694.1 | 1172 | 132 | |||
| XM_028845862.1 | XP_028701695.1 | 1073 | 132 | |||
| XM_028845863.1 | XP_028701696.1 | 953 | 132 | |||
| GenBank mRNA | JU485860.1 | - | 650 | 132 | ||
More Information
Set Membership: |
|

GPR141
Genomic Map