Assay Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135276.2 | XP_014990762.2 | 255 | 75 | ||
| XM_028841105.1 | XP_028696938.1 | 280 | 75 | |||
| XM_028841110.1 | XP_028696943.1 | 280 | 75 | |||
| GenBank mRNA | JU481989.1 | - | 149 | 75 | ||
| JV644650.1 | - | 213 | 75 | |||
More Information
Set Membership: |
|

FAAP20
Genomic Map