Assay Details
| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194623.1 | NP_001181552.1 | 754 | 131 | ||
| XM_015124385.2 | XP_014979871.1 | 1426 | 131 | |||
| XM_015124386.2 | XP_014979872.1 | 995 | 131 | |||
| XM_015124387.2 | XP_014979873.1 | 1345 | 131 | |||
More Information
Set Membership: |
|

IZUMO1
Genomic Map