Assay Details
| Gene Symbol: VSIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092052.4 | XP_001092052.2 | 240 | 69 | ||
| XM_015127970.2 | XP_014983456.1 | 235 | 69 | |||
| XM_028842173.1 | XP_028698006.1 | 137 | 69 | |||
| XM_028842174.1 | XP_028698007.1 | 233 | 69 | |||
| GenBank mRNA | JV045735.1 | - | 209 | 69 | ||
| JV635434.1 | - | 197 | 69 | |||
| JV636809.1 | - | 197 | 69 | |||
More Information
Set Membership: |
|

VSIG1
Genomic Map