Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133440.2 | XP_014988926.1 | 1167 | 69 | ||
| XM_015133441.2 | XP_014988927.1 | 1149 | 69 | |||
| XM_028845736.1 | XP_028701569.1 | 1407 | 69 | |||
| XM_028845737.1 | XP_028701570.1 | 1425 | 69 | |||
| GenBank mRNA | JU334352.1 | - | 199 | 69 | ||
| JU473194.1 | - | 198 | 69 | |||
More Information
Set Membership: |
|

ZNF713
Genomic Map