Assay Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257485.1 | NP_001244414.1 | 1120 | 88 | ||
| GenBank mRNA | JU335970.1 | - | 1120 | 88 | ||
| JU476853.1 | - | 1109 | 88 | |||
| JV047965.1 | - | 1105 | 88 | |||
More Information
Set Membership: |
|

MRPS9
Genomic Map