Assay Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091410.4 | XP_001091410.3 | 793 | 78 | ||
| XM_015137032.2 | XP_014992518.1 | 793 | 78 | |||
| XM_015137033.2 | XP_014992519.1 | 580 | 78 | |||
| XM_015137036.2 | XP_014992522.1 | 793 | 78 | |||
| XM_028847300.1 | XP_028703133.1 | 949 | 78 | |||
| XM_028847301.1 | XP_028703134.1 | 755 | 78 | |||
| GenBank mRNA | JU471983.1 | - | 538 | 78 | ||
| JU471984.1 | - | 538 | 78 | |||
| JU471985.1 | - | 538 | 78 | |||
More Information
Set Membership: |
|

SCML4
Genomic Map