Assay Details
Gene Symbol: C19H19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194075.3 | NP_001181004.2 | 326 | 134 | ||
XM_015123960.2 | XP_014979446.2 | 1247 | 134 | |||
XM_028838387.1 | XP_028694220.1 | 667 | 134 | |||
XM_028838388.1 | XP_028694221.1 | 650 | 134 | |||
XM_028838389.1 | XP_028694222.1 | 117 | 134 | |||
XM_028838390.1 | XP_028694223.1 | 1247 | 134 | |||
XM_028838391.1 | XP_028694224.1 | 117 | 134 | |||
XM_028838392.1 | XP_028694225.1 | 667 | 134 | |||
XM_028838393.1 | XP_028694226.1 | 650 | 134 | |||
GenBank mRNA | JV636107.1 | - | 446 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028838387.1, XM_028838388.1, XM_028838389.1, XM_028838391.1, XM_028838392.1, XM_028838393.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
