Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126729.2 | XP_014982215.2 | 193 | 134 | ||
| XM_015126730.2 | XP_014982216.2 | 192 | 134 | |||
| XM_015126731.2 | XP_014982217.2 | 192 | 134 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015126729.2, XM_015126730.2, XM_015126731.2 transcripts.
Set Membership: |
|

CIBAR2
Genomic Map