Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126729.2 | XP_014982215.2 | 425 | 184 | ||
| XM_015126731.2 | XP_014982217.2 | 424 | 184 | |||
More Information
Set Membership: |
|

CIBAR2
Genomic Map