Assay Details
| Gene Symbol: VWA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091779.4 | XP_001091779.1 | 380 | 86 | ||
| XM_015148229.2 | XP_015003715.2 | 380 | 86 | |||
| XM_015148230.2 | XP_015003716.2 | 493 | 86 | |||
| XM_015148231.2 | XP_015003717.2 | 272 | 86 | |||
More Information
Set Membership: |
|

VWA2
Genomic Map