Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122247.2 | XP_014977733.2 | 1836 | 135 | ||
| XM_028839118.1 | XP_028694951.1 | 603 | 135 | |||
More Information
Set Membership: |
|

SPATA21
Genomic Map